Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col6a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB370265.1 | - | 33 - 34 | 6025 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |