Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qrich1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114119.1 | NP_001107591.1 | 7 - 8 | 2087 | 95 | |
NM_175143.5 | NP_780352.2 | 6 - 7 | 1963 | 95 | ||
GenBank mRNA | AK151491.1 | - | 7 - 8 | 2061 | 95 | |
AK151530.1 | - | 7 - 8 | 2061 | 95 | ||
BC006738.1 | - | 6 - 7 | 1940 | 3156574 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |