Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171052.1 | NP_001164523.1 | 7 - 8 | 699 | 63 | |
NM_001171053.1 | NP_001164524.1 | 7 - 8 | 699 | 63 | ||
NM_001171054.1 | NP_001164525.1 | 7 - 8 | 699 | 63 | ||
NM_054082.2 | NP_473423.1 | 7 - 8 | 696 | 63 | ||
GenBank mRNA | AF288138.1 | - | 7 - 8 | 650 | 63 | |
AK129318.1 | - | 7 - 8 | 628 | 63 | ||
AK145942.1 | - | 7 - 8 | 644 | 63 | ||
AK146721.1 | - | 7 - 8 | 652 | 63 | ||
AK146905.1 | - | 7 - 8 | 652 | 63 | ||
AK147197.1 | - | 7 - 8 | 642 | 63 | ||
AK154856.1 | - | 7 - 8 | 639 | 63 | ||
AK158866.1 | - | 6 - 7 | 562 | 63 | ||
BC022124.1 | - | 7 - 8 | 663 | 5097283 | 63 | |
BC099960.1 | - | 7 - 8 | 616 | 30023887 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |