Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700010055.mm.1 |
NM_001037997.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010055.mm.1 |
NM_001286415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010055.mm.1 |
TC1700001053.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010054.mm.1 |
NM_001037997.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010054.mm.1 |
NM_001286415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010054.mm.1 |
TC1700001053.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010050.mm.1 |
NM_001037997.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010050.mm.1 |
TC1700001053.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010051.mm.1 |
NM_001286415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010051.mm.1 |
TC1700001053.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010053.mm.1 |
NM_001037997.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010053.mm.1 |
TC1700001053.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010056.mm.1 |
NM_001037997.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010056.mm.1 |
NM_001286415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010056.mm.1 |
TC1700001053.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010057.mm.1 |
NM_001037997.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010057.mm.1 |
NM_001286415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010057.mm.1 |
TC1700001053.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010058.mm.1 |
NM_001037997.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010058.mm.1 |
NM_001286415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010058.mm.1 |
TC1700001053.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010052.mm.1 |
NM_001037997.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010052.mm.1 |
NM_001286415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010052.mm.1 |
TC1700001053.mm.1 |
MTA TC |