Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081122.1 | NP_001074591.1 | 1 - 2 | 221 | 72 | |
NM_001301689.1 | NP_001288618.1 | 1 - 2 | 221 | 72 | ||
GenBank mRNA | AK076549.1 | - | 1 - 2 | 222 | 72 | |
KC211185.1 | - | 1 - 2 | 78 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |