Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081287.2 | NP_001074756.2 | 2 - 3 | 272 | 60 | |
NM_001161620.1 | NP_001155092.1 | 2 - 3 | 272 | 60 | ||
GenBank mRNA | AK017344.1 | - | 5 - 6 | 634 | 60 | |
AK033081.1 | - | 2 - 3 | 232 | 60 | ||
AK035303.1 | - | 2 - 3 | 264 | 60 | ||
AK036956.1 | - | 2 - 3 | 243 | 60 | ||
AK050710.1 | - | 2 - 3 | 269 | 60 | ||
AK078849.1 | - | 2 - 3 | 271 | 60 | ||
AK162694.1 | - | 2 - 3 | 272 | 60 | ||
BC049662.1 | - | 2 - 3 | 226 | 60 | ||
BC117550.1 | - | 1 - 2 | 119 | 40105182 | 60 | |
BC118058.1 | - | 1 - 2 | 119 | 40105180 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |