Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029896.1 | NP_084172.1 | 3 - 4 | 616 | 132 | |
GenBank mRNA | AK035109.1 | - | 3 - 4 | 618 | 132 | |
AK037620.1 | - | 3 - 4 | 623 | 132 | ||
AK149692.1 | - | 3 - 4 | 617 | 132 | ||
AK159240.1 | - | 3 - 4 | 616 | 131 | ||
BC019115.1 | - | 2 - 3 | 142 | 132 | ||
BC031502.1 | - | 3 - 4 | 306 | 132 | ||
BC094419.1 | - | 3 - 4 | 579 | 132 | ||
BC157943.1 | - | 539 | 9007278 | 132 | ||
BC157945.1 | - | 540 | 9007280 | 132 | ||
BC158054.1 | - | 540 | 9007356 | 132 | ||
EF011615.1 | - | 3 - 4 | 305 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |