Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252467.1 | NP_001239396.1 | 2 - 3 | 247 | 109 | |
NM_027933.3 | NP_082209.1 | 2 - 3 | 247 | 109 | ||
NR_045519.1 | 2 - 3 | 247 | 109 | |||
GenBank mRNA | AK133949.1 | - | 2 - 3 | 94 | 109 | |
AK154201.1 | - | 2 - 3 | 106 | 109 | ||
AK155301.1 | - | 2 - 3 | 116 | 109 | ||
AK167731.1 | - | 2 - 3 | 97 | 109 | ||
AK183251.1 | - | 78 | 109 | |||
BB842976.1 | - | 2 - 3 | 245 | 109 | ||
BC066005.1 | - | 2 - 3 | 88 | 109 | ||
BC145892.1 | - | 2 - 3 | 111 | 40131195 | 109 | |
BC145894.1 | - | 2 - 3 | 111 | 40131197 | 109 | |
CN717165.1 | - | 2 - 3 | 178 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |