Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017525.1 | NP_001017525.1 | 12 - 13 | 1594 | 71 | |
NM_028709.2 | NP_082985.2 | 14 - 15 | 3447 | 71 | ||
GenBank mRNA | AK018115.1 | - | 4 - 5 | 908 | 71 | |
AK039038.1 | - | 11 - 12 | 1390 | 71 | ||
AK044022.1 | - | 12 - 13 | 1485 | 71 | ||
BC059036.1 | - | 2 - 3 | 465 | 71 | ||
BC072592.1 | - | 14 - 15 | 2072 | 71 | ||
BC096535.1 | - | 14 - 15 | 3447 | 71 | ||
BC141145.1 | - | 14 - 15 | 2519 | 9055757 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |