Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252478.1 | NP_001239407.1 | 11 - 12 | 838 | 92 | |
NM_144798.6 | NP_659047.2 | 11 - 12 | 823 | 92 | ||
GenBank mRNA | AF233346.1 | - | 10 - 11 | 775 | 92 | |
AF395840.1 | - | 10 - 11 | 765 | 92 | ||
AK082907.1 | - | 11 - 12 | 800 | 92 | ||
AK150342.1 | - | 11 - 12 | 799 | 92 | ||
AK151120.1 | - | 11 - 12 | 799 | 92 | ||
AK160903.1 | - | 11 - 12 | 827 | 92 | ||
BC005753.1 | - | 3 - 4 | 286 | 92 | ||
BC066162.1 | - | 11 - 12 | 790 | 30471027 | 92 | |
BC068169.1 | - | 11 - 12 | 812 | 30542277 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |