Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007771.3 | NP_031797.1 | 11 - 12 | 2301 | 140 | |
GenBank mRNA | AB000777.1 | - | 11 - 12 | 1737 | 140 | |
AF156986.1 | - | 11 - 12 | 2036 | 140 | ||
AK133818.1 | - | 11 - 12 | 2313 | 140 | ||
AK162460.1 | - | 11 - 12 | 2301 | 140 | ||
BC022174.1 | - | 11 - 12 | 2072 | 4036758 | 140 | |
BC085499.1 | - | 11 - 12 | 2301 | 6813367 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |