Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700011706.mm.1 |
NM_001159496.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011706.mm.1 |
NM_001159496.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011706.mm.1 |
NM_001159497.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011706.mm.1 |
NM_001159497.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011706.mm.1 |
NM_001159498.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011706.mm.1 |
NM_001159498.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011706.mm.1 |
NM_011151.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011706.mm.1 |
NM_011151.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011706.mm.1 |
TC1700001266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700011703.mm.1 |
NM_001159498.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011703.mm.1 |
NM_001159498.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011703.mm.1 |
TC1700001266.mm.1 |
MTA TC |