Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010214.4 | NP_034344.2 | 1 - 2 | 194 | 81 | |
GenBank mRNA | AK006464.1 | - | 1 - 2 | 165 | 81 | |
AK029585.1 | - | 195 | 81 | |||
AK161287.1 | - | 1 - 2 | 172 | 81 | ||
BC061148.1 | - | 1 - 2 | 141 | 6774577 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |