Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aff3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290814.1 | NP_001277743.1 | 12 - 13 | 2572 | 82 | |
GenBank mRNA | AK014322.1 | - | 9 - 10 | 1879 | 82 | |
AK209098.1 | - | 1 - 2 | 119 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |