Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033367.3 | NP_001028539.1 | 6 - 7 | 2738 | 107 | |
GenBank mRNA | AK035300.1 | - | 3 - 4 | 2139 | 107 | |
AK143864.1 | - | 6 - 7 | 2739 | 107 | ||
AK171624.1 | - | 6 - 7 | 2733 | 107 | ||
BC144861.1 | - | 4 - 5 | 2509 | 107 | ||
BC144862.1 | - | 4 - 5 | 2495 | 107 | ||
BC145617.1 | - | 5 - 6 | 2532 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |