Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153552.3 | NP_705780.1 | 4 - 5 | 264 | 82 | |
GenBank mRNA | AK031785.1 | - | 4 - 5 | 265 | 82 | |
AK032200.1 | - | 4 - 5 | 265 | 82 | ||
AK042867.1 | - | 4 - 5 | 270 | 82 | ||
AK052835.1 | - | 4 - 5 | 259 | 82 | ||
AK139307.1 | - | 4 - 5 | 270 | 82 | ||
AK167374.1 | - | 4 - 5 | 278 | 82 | ||
AK168619.1 | - | 4 - 5 | 529 | 82 | ||
BC024951.1 | - | 4 - 5 | 436 | 4010605 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |