Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029402.3 | NP_083678.2 | 16 - 17 | 1808 | 75 | |
GenBank mRNA | AK004875.2 | - | 16 - 17 | 1764 | 75 | |
AK016520.1 | - | 16 - 17 | 1778 | 75 | ||
AK028589.1 | - | 16 - 17 | 1777 | 75 | ||
AK144693.1 | - | 16 - 17 | 1782 | 75 | ||
AK160597.1 | - | 16 - 17 | 1793 | 75 | ||
AK220287.1 | - | 16 - 17 | 1783 | 75 | ||
BC025902.1 | - | 8 - 9 | 874 | 75 | ||
BC026779.1 | - | 16 - 17 | 1709 | 4018298 | 75 | |
BC027428.1 | - | 16 - 17 | 1771 | 3983461 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |