Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029402.3 | NP_083678.2 | 7 - 8 | 801 | 136 | |
GenBank mRNA | AK004875.2 | - | 7 - 8 | 757 | 136 | |
AK016520.1 | - | 7 - 8 | 771 | 136 | ||
AK028589.1 | - | 7 - 8 | 770 | 136 | ||
AK144693.1 | - | 7 - 8 | 774 | 136 | ||
AK160597.1 | - | 7 - 8 | 786 | 136 | ||
AK220287.1 | - | 7 - 8 | 776 | 136 | ||
BC026779.1 | - | 7 - 8 | 702 | 4018298 | 136 | |
BC027428.1 | - | 7 - 8 | 764 | 3983461 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |