Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190409.1 | NP_001177338.1 | 7 - 8 | 966 | 84 | |
NM_025952.4 | NP_080228.4 | 7 - 8 | 966 | 84 | ||
GenBank mRNA | AK010320.1 | - | 7 - 8 | 902 | 84 | |
AK012185.1 | - | 7 - 8 | 843 | 84 | ||
AK013243.1 | - | 7 - 8 | 847 | 84 | ||
AK018623.1 | - | 7 - 8 | 869 | 84 | ||
AK136622.1 | - | 7 - 8 | 848 | 84 | ||
AK144696.1 | - | 7 - 8 | 845 | 84 | ||
BC003881.1 | - | 7 - 8 | 853 | 3496572 | 84 | |
DQ000005.1 | - | 7 - 8 | 966 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |