Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009985.4 | NP_034115.2 | 3 - 4 | 289 | 69 | |
GenBank mRNA | AF014941.1 | - | 3 - 4 | 274 | 69 | |
AK088375.1 | - | 3 - 4 | 290 | 69 | ||
BB850740.1 | - | 3 - 4 | 290 | 69 | ||
BC019136.1 | - | 3 - 4 | 290 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |