Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sipa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164480.1 | NP_001157952.1 | 13 - 14 | 3001 | 65 | |
NM_001164481.1 | NP_001157953.1 | 14 - 15 | 3255 | 65 | ||
NM_001164482.1 | NP_001157954.1 | 12 - 13 | 2940 | 65 | ||
NM_001164568.1 | NP_001158040.1 | 12 - 13 | 2970 | 65 | ||
NM_011379.4 | NP_035509.4 | 13 - 14 | 3224 | 65 | ||
GenBank mRNA | AK028151.1 | - | 12 - 13 | 2211 | 65 | |
AK137578.1 | - | 13 - 14 | 2947 | 65 | ||
AK149669.1 | - | 13 - 14 | 2947 | 65 | ||
AK156280.1 | - | 14 - 15 | 3209 | 65 | ||
AK169945.1 | - | 12 - 13 | 2886 | 65 | ||
AK220472.1 | - | 13 - 14 | 3137 | 65 | ||
BC043679.1 | - | 5 - 6 | 1361 | 65 | ||
BC054824.1 | - | 13 - 14 | 2945 | 5324326 | 65 | |
D11374.1 | - | 12 - 13 | 2898 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |