Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013722.3 | NP_038750.2 | 8 - 9 | 1168 | 69 | |
GenBank mRNA | AF192754.1 | - | 8 - 9 | 1113 | 69 | |
AF498252.1 | - | 7 - 8 | 915 | 69 | ||
AF498253.1 | - | 7 - 8 | 915 | 69 | ||
AK048643.1 | - | 8 - 9 | 1206 | 69 | ||
AK080557.1 | - | 8 - 9 | 1130 | 69 | ||
BC107397.2 | - | 7 - 8 | 1034 | 40058225 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |