Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700009994.mm.1 |
NM_001081653.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009994.mm.1 |
NM_001081653.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009994.mm.1 |
TC1700001036.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700009996.mm.1 |
NM_001081653.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009996.mm.1 |
NM_001081653.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009996.mm.1 |
TC1700001036.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010005.mm.1 |
NM_001081653.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010005.mm.1 |
NM_001081653.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010005.mm.1 |
TC1700001036.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700009991.mm.1 |
NM_001081653.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009991.mm.1 |
NM_001081653.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009991.mm.1 |
TC1700001036.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700009997.mm.1 |
NM_001081653.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009997.mm.1 |
NM_001081653.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009997.mm.1 |
TC1700001036.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700009999.mm.1 |
NM_001081653.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009999.mm.1 |
NM_001081653.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009999.mm.1 |
TC1700001036.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010000.mm.1 |
NM_001081653.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010000.mm.1 |
NM_001081653.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010000.mm.1 |
TC1700001036.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010004.mm.1 |
NM_001081653.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010004.mm.1 |
NM_001081653.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010004.mm.1 |
TC1700001036.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700009992.mm.1 |
NM_001081653.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009992.mm.1 |
NM_001081653.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009992.mm.1 |
TC1700001036.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010001.mm.1 |
NM_001081653.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010001.mm.1 |
NM_001081653.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010001.mm.1 |
TC1700001036.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010008.mm.1 |
NM_001081653.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010008.mm.1 |
NM_001081653.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010008.mm.1 |
TC1700001036.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700009995.mm.1 |
NM_001081653.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009995.mm.1 |
NM_001081653.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009995.mm.1 |
TC1700001036.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010003.mm.1 |
NM_001081653.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010003.mm.1 |
NM_001081653.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010003.mm.1 |
TC1700001036.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700009998.mm.1 |
NM_001081653.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009998.mm.1 |
NM_001081653.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009998.mm.1 |
TC1700001036.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010002.mm.1 |
NM_001081653.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010002.mm.1 |
NM_001081653.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010002.mm.1 |
TC1700001036.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010007.mm.1 |
NM_001081653.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010007.mm.1 |
NM_001081653.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010007.mm.1 |
TC1700001036.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700009993.mm.1 |
NM_001081653.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009993.mm.1 |
NM_001081653.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700009993.mm.1 |
TC1700001036.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700010006.mm.1 |
NM_001081653.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010006.mm.1 |
NM_001081653.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700010006.mm.1 |
TC1700001036.mm.1 |
MTA TC |