Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1rl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025602.3 | NP_001020773.1 | 2 - 3 | 430 | 92 | |
NM_001294171.1 | NP_001281100.1 | 2 - 3 | 263 | 92 | ||
NM_010743.3 | NP_034873.2 | 2 - 3 | 430 | 92 | ||
GenBank mRNA | AK033417.1 | - | 2 - 3 | 283 | 92 | |
BC051445.1 | - | 2 - 3 | 240 | 30073421 | 92 | |
D13695.1 | - | 1 - 2 | 122 | 92 | ||
M24843.1 | - | 2 - 3 | 271 | 92 | ||
Y07519.1 | - | 2 - 3 | 274 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |