Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000008748.mm.1 |
NM_001168684.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008748.mm.1 |
NM_001168684.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008748.mm.1 |
NM_172051.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008748.mm.1 |
NM_172051.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008748.mm.1 |
NM_177026.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008748.mm.1 |
NM_177026.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008748.mm.1 |
TC1000001149.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008747.mm.1 |
NM_001168684.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008747.mm.1 |
NM_001168684.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008747.mm.1 |
NM_172051.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008747.mm.1 |
NM_172051.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008747.mm.1 |
NM_177026.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008747.mm.1 |
NM_177026.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008747.mm.1 |
TC1000001149.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008746.mm.1 |
NM_001168684.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008746.mm.1 |
NM_001168684.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008746.mm.1 |
NM_172051.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008746.mm.1 |
NM_172051.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008746.mm.1 |
NM_177026.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008746.mm.1 |
NM_177026.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008746.mm.1 |
TC1000001149.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008743.mm.1 |
NM_001168684.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008743.mm.1 |
NM_001168684.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008743.mm.1 |
TC1000001149.mm.1 |
MTA TC |