Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000009281.mm.1 |
NM_001162368.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009281.mm.1 |
NM_001162368.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009281.mm.1 |
NM_001162369.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009281.mm.1 |
NM_001162369.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009281.mm.1 |
NM_001205098.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009281.mm.1 |
NM_001205098.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009281.mm.1 |
TC1000001228.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000009266.mm.1 |
NM_001162368.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009266.mm.1 |
NM_001162368.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009266.mm.1 |
TC1000001228.mm.1 |
MTA TC |