Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162904.1 | NP_001156376.1 | 3 - 4 | 667 | 78 | |
NM_010785.2 | NP_034915.2 | 3 - 4 | 667 | 78 | ||
NM_148922.3 | NP_683724.2 | 3 - 4 | 667 | 78 | ||
GenBank mRNA | AK004789.2 | - | 3 - 4 | 622 | 78 | |
AK011769.1 | - | 3 - 4 | 622 | 78 | ||
BC052680.1 | - | 3 - 4 | 617 | 78 | ||
BC137608.1 | - | 3 - 4 | 595 | 8860627 | 78 | |
BC144777.1 | - | 3 - 4 | 595 | 9053325 | 78 | |
EU490427.1 | - | 3 - 4 | 509 | 78 | ||
M20823.1 | - | 3 - 4 | 530 | 78 | ||
M20824.1 | - | 3 - 4 | 530 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |