Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162904.1 | NP_001156376.1 | 4 - 5 | 802 | 71 | |
NM_010785.2 | NP_034915.2 | 4 - 5 | 802 | 71 | ||
NM_148922.3 | NP_683724.2 | 4 - 5 | 802 | 71 | ||
GenBank mRNA | AK004789.2 | - | 4 - 5 | 757 | 71 | |
AK011769.1 | - | 4 - 5 | 757 | 71 | ||
BC052680.1 | - | 4 - 5 | 752 | 71 | ||
BC137608.1 | - | 4 - 5 | 730 | 8860627 | 71 | |
BC144777.1 | - | 4 - 5 | 730 | 9053325 | 71 | |
EU490427.1 | - | 4 - 5 | 644 | 71 | ||
M20823.1 | - | 4 - 5 | 665 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |