Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007935.2 | NP_031961.1 | 4 - 5 | 760 | 90 | |
NM_027497.3 | NP_081773.1 | 4 - 5 | 911 | 90 | ||
GenBank mRNA | AF079765.1 | - | 4 - 5 | 739 | 90 | |
AK040254.1 | - | 4 - 5 | 911 | 90 | ||
AK148194.1 | - | 4 - 5 | 729 | 90 | ||
AK149317.1 | - | 1 - 2 | 111 | 90 | ||
BC138622.1 | - | 4 - 5 | 739 | 8861642 | 90 | |
BC138623.1 | - | 4 - 5 | 739 | 8861643 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |