Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdpcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145425.3 | NP_663400.2 | 16 - 17 | 2202 | 140 | |
GenBank mRNA | AF424701.1 | - | 16 - 17 | 2165 | 140 | |
AK043672.1 | - | 15 - 16 | 2089 | 140 | ||
AK083042.1 | - | 16 - 17 | 2203 | 140 | ||
BC141166.1 | - | 16 - 17 | 2166 | 9055778 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |