Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdpcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145425.3 | NP_663400.2 | 4 - 5 | 269 | 116 | |
GenBank mRNA | AF424701.1 | - | 4 - 5 | 232 | 116 | |
AK043672.1 | - | 3 - 4 | 156 | 116 | ||
AK082370.1 | - | 4 - 5 | 245 | 116 | ||
AK083042.1 | - | 4 - 5 | 270 | 116 | ||
BC141166.1 | - | 4 - 5 | 233 | 9055778 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |