Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252515.1 | NP_001239444.1 | 13 - 14 | 1946 | 144 | |
NM_153078.4 | NP_694718.4 | 13 - 14 | 1871 | 144 | ||
GenBank mRNA | AF424697.1 | - | 12 - 13 | 1721 | 144 | |
AK173055.1 | - | 13 - 14 | 1947 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |