Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pus10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033654.2 | NP_001028826.1 | 3 - 4 | 453 | 116 | |
NM_028304.2 | NP_082580.1 | 3 - 4 | 866 | 116 | ||
NM_028956.4 | NP_083232.1 | 3 - 4 | 463 | 116 | ||
GenBank mRNA | AK012737.1 | - | 3 - 4 | 630 | 116 | |
AK017063.1 | - | 3 - 4 | 440 | 116 | ||
AK028368.1 | - | 3 - 4 | 450 | 116 | ||
AK029866.1 | - | 3 - 4 | 454 | 116 | ||
AK088562.1 | - | 3 - 4 | 454 | 116 | ||
AK145055.1 | - | 3 - 4 | 812 | 116 | ||
BC025555.1 | - | 3 - 4 | 684 | 5321811 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |