Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159290.1 | NP_001152762.1 | 1 - 2 | 44 | 75 | |
GenBank mRNA | AK045556.1 | - | 1 - 2 | 45 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |