Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggact | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145466.2 | NP_663441.1 | 1 - 2 | 66 | 107 | |
GenBank mRNA | AK050251.1 | - | 1 - 2 | 56 | 107 | |
BC006662.1 | - | 1 - 2 | 3501534 | 107 | ||
BY119987.1 | - | 1 - 2 | 64 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |