Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00017955.mm.1 |
NM_001001176.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017955.mm.1 |
NM_001001176.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017955.mm.1 |
NM_001167988.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017955.mm.1 |
NM_001167988.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017955.mm.1 |
TC0X00002826.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00017956.mm.1 |
NM_001001176.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017956.mm.1 |
NM_001001176.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017956.mm.1 |
NM_001167988.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017956.mm.1 |
NM_001167988.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017956.mm.1 |
TC0X00002826.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00017957.mm.1 |
NM_001001176.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017957.mm.1 |
NM_001001176.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017957.mm.1 |
NM_001167988.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017957.mm.1 |
NM_001167988.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017957.mm.1 |
TC0X00002826.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00017954.mm.1 |
NM_001001176.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017954.mm.1 |
NM_001001176.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017954.mm.1 |
NM_001167988.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017954.mm.1 |
NM_001167988.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017954.mm.1 |
TC0X00002826.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00017958.mm.1 |
NM_001001176.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017958.mm.1 |
NM_001001176.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017958.mm.1 |
TC0X00002826.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00017961.mm.1 |
NM_001167988.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017961.mm.1 |
NM_001167988.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017961.mm.1 |
TC0X00002826.mm.1 |
MTA TC |