Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026241.3 | NP_080517.1 | 2 - 3 | 218 | 72 | |
GenBank mRNA | AK018681.1 | - | 2 - 3 | 219 | 72 | |
AK030874.1 | - | 414 | 72 | |||
AK042896.1 | - | 2 - 3 | 219 | 72 | ||
AK081207.1 | - | 2 - 3 | 411 | 72 | ||
AK158215.1 | - | 2 - 3 | 413 | 72 | ||
AK160231.1 | - | 2 - 3 | 234 | 72 | ||
BC117004.1 | - | 2 - 3 | 248 | 40126323 | 72 | |
BC119100.1 | - | 2 - 3 | 248 | 8733849 | 72 | |
BC144893.1 | - | 2 - 3 | 248 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |