Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026850.4 | NP_081126.2 | 2 - 3 | 164 | 149 | |
GenBank mRNA | AF110512.1 | - | 1 - 2 | 134 | 149 | |
AK004334.1 | - | 2 - 3 | 162 | 149 | ||
AK078372.1 | - | 2 - 3 | 165 | 149 | ||
AK168509.1 | - | 2 - 3 | 205 | 149 | ||
BC005601.1 | - | 2 - 3 | 185 | 3483084 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |