Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177772.4 | NP_808440.2 | 4 - 5 | 453 | 99 | |
GenBank mRNA | AK028764.1 | - | 4 - 5 | 444 | 99 | |
AK132518.1 | - | 4 - 5 | 489 | 99 | ||
BC112376.1 | - | 4 - 5 | 441 | 40057331 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |