Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930594M22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK019841.1 | - | 1 - 2 | 220 | 67 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | gamma-glutamylamine cyclotransferase | Ggact | A2ld1 | Anti-sense | Chr. 14 - 122890860 - 122913165 on NCBI Build GRCm38 | |
Downstream | predicted gene 15735 | Gm15735 | OTTMUSG00000025808 | 18.0kb | Sense | Chr. 14 - 122950510 - 122956357 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons |