Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009592.1 | NP_033722.1 | 1 - 2 | 195 | 95 | |
GenBank mRNA | AK040069.1 | - | 1 - 2 | 193 | 95 | |
AK084853.1 | - | 1 - 2 | 186 | 95 | ||
AK151967.1 | - | 1 - 2 | 181 | 95 | ||
AK152535.1 | - | 1 - 2 | 181 | 95 | ||
AK152816.1 | - | 1 - 2 | 181 | 95 | ||
AK152946.1 | - | 1 - 2 | 182 | 95 | ||
BB612154.1 | - | 1 - 2 | 195 | 95 | ||
BC080769.1 | - | 1 - 2 | 217 | 95 | ||
BC151037.1 | - | 1 - 2 | 195 | 9087950 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons |