Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkdcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134117.2 | NP_598878.2 | 1 - 2 | 822 | 89 | |
GenBank mRNA | AB381893.1 | - | 1 - 2 | 814 | 89 | |
AB436780.1 | - | 1 - 2 | 803 | 89 | ||
BC086639.1 | - | 2 - 3 | 660 | 6828629 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |