Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkdcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134117.2 | NP_598878.2 | 5 - 6 | 1402 | 83 | |
GenBank mRNA | AB381893.1 | - | 5 - 6 | 1394 | 83 | |
AB436780.1 | - | 5 - 6 | 1383 | 83 | ||
AK133913.1 | - | 1 - 2 | 79 | 83 | ||
BC022157.1 | - | 4 - 5 | 549 | 83 | ||
BC086639.1 | - | 6 - 7 | 1240 | 6828629 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |