Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync2li1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172256.1 | NP_758460.1 | 3 - 4 | 238 | 116 | |
GenBank mRNA | AK008822.1 | - | 2 - 3 | 188 | 116 | |
AK160259.1 | - | 3 - 4 | 252 | 116 | ||
BC030450.1 | - | 3 - 4 | 240 | 5400492 | 116 | |
BC039070.1 | - | 3 - 4 | 238 | 3482503 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |