Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nckipsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030729.4 | NP_109654.2 | 4 - 5 | 685 | 75 | |
GenBank mRNA | AF130313.2 | - | 4 - 5 | 581 | 75 | |
AK134725.1 | - | 4 - 5 | 667 | 75 | ||
BC064818.1 | - | 4 - 5 | 685 | 30292108 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |