Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011790.4 | NP_035920.1 | 11 - 12 | 1261 | 71 | |
GenBank mRNA | AF124664.1 | - | 10 - 11 | 1088 | 71 | |
AJ130975.1 | - | 10 - 11 | 1120 | 71 | ||
AK011094.1 | - | 11 - 12 | 1268 | 71 | ||
AK045012.1 | - | 1244 | 71 | |||
AK149936.1 | - | 11 - 12 | 1241 | 71 | ||
AK167104.1 | - | 11 - 12 | 1218 | 71 | ||
AK168055.1 | - | 11 - 12 | 1266 | 71 | ||
BC051998.1 | - | 11 - 12 | 1261 | 6400287 | 71 | |
BC052422.1 | - | 11 - 12 | 1261 | 6400287 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |