Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028719.1 | NP_082995.1 | 7 - 8 | 997 | 107 | |
GenBank mRNA | AK043692.1 | - | 7 - 8 | 1010 | 107 | |
BC043087.1 | - | 7 - 8 | 997 | 6417611 | 107 | |
BC063081.1 | - | 7 - 8 | 997 | 6417611 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |