Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027354.2 | NP_081630.2 | 3 - 4 | 651 | 67 | |
GenBank mRNA | AK011064.1 | - | 3 - 4 | 329 | 67 | |
AK154163.1 | - | 3 - 4 | 401 | 67 | ||
BC034901.1 | - | 3 - 4 | 311 | 5255069 | 67 | |
CA557124.1 | - | 3 - 4 | 301 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |