Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053268.2 | NP_444498.2 | 5 - 6 | 527 | 138 | |
GenBank mRNA | AB056433.1 | - | 5 - 6 | 520 | 138 | |
AK034434.1 | - | 5 - 6 | 527 | 138 | ||
AK154499.1 | - | 5 - 6 | 544 | 138 | ||
AK166654.1 | - | 5 - 6 | 527 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |