Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018818.2 | NP_061288.2 | 2 - 3 | 138 | 78 | |
GenBank mRNA | AF218084.1 | - | 2 - 3 | 138 | 78 | |
AK021238.1 | - | 2 - 3 | 141 | 78 | ||
AK035956.1 | - | 2 - 3 | 133 | 78 | ||
AK141830.1 | - | 2 - 3 | 133 | 78 | ||
BC047461.1 | - | 2 - 3 | 128 | 5067077 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |